Assay Details
Gene Symbol: CCND3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034709.2 | NP_001029881.1 | 2 - 3 | 500 | 78 | |
GenBank mRNA | BC105236.1 | - | 2 - 3 | 501 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
