Assay Details
Gene Symbol: CCND3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034709.2 | NP_001029881.1 | 3 - 4 | 664 | 97 | |
GenBank mRNA | BC105236.1 | - | 3 - 4 | 665 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
