Assay Details
| Gene Symbol: RPS10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034716.2 | NP_001029888.1 | 4 - 5 | 435 | 63 | |
| GenBank mRNA | AB098806.1 | - | 2 - 3 | 146 | 63 | |
| BC102416.1 | - | 4 - 5 | 438 | 63 | ||
More Information
Set Membership: |
|

RPS10
Genomic Map