Assay Details
| Gene Symbol: RAMAC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034757.1 | NP_001029929.1 | 4 | 951 | 106 | |
| GenBank mRNA | BC102563.1 | - | 4 | 951 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAMAC| Gene Symbol: RAMAC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034757.1 | NP_001029929.1 | 4 | 951 | 106 | |
| GenBank mRNA | BC102563.1 | - | 4 | 951 | 106 | |
Set Membership: |
|