Assay Details
| Gene Symbol: RPL32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034783.2 | NP_001029955.1 | 1 - 2 | 46 | 98 | |
| GenBank mRNA | BC102748.1 | - | 1 - 2 | 47 | 98 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102748.1, NM_001034783.2 transcripts.
Set Membership: |
|

RPL32
Genomic Map