Assay Details
| Gene Symbol: RPL32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034783.2 | NP_001029955.1 | 3 - 4 | 322 | 95 | |
| GenBank mRNA | BC102748.1 | - | 3 - 4 | 323 | 95 | |
More Information
Set Membership: |
|

RPL32
Genomic Map