Assay Details
| Gene Symbol: UQCRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034797.2 | NP_001029969.1 | 3 - 4 | 270 | 66 | |
| GenBank mRNA | BC103057.1 | - | 3 - 4 | 271 | 66 | |
More Information
Set Membership: |
|

UQCRB
Genomic Map