Assay Details
| Gene Symbol: APOC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034807.1 | NP_001029979.1 | 1 - 2 | 111 | 64 | |
| GenBank mRNA | BC103430.1 | - | 1 - 2 | 111 | 64 | |
More Information
Set Membership: |
|

APOC4
Genomic Map