Assay Details
| Gene Symbol: RPL37A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035008.2 | NP_001030180.1 | 1 - 2 | 28 | 63 | |
| GenBank mRNA | BC102044.1 | - | 1 - 2 | 29 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102044.1, NM_001035008.2 transcripts.
Set Membership: |
|

RPL37A
Genomic Map