Assay Details
| Gene Symbol: RAB19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035040.2 | NP_001030212.1 | 1 - 2 | 55 | 80 | |
| GenBank mRNA | BC102961.1 | - | 1 - 2 | 58 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102961.1, NM_001035040.2 transcripts.
Set Membership: |
|

RAB19
Genomic Map