Assay Details
Gene Symbol: RAB19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035040.2 | NP_001030212.1 | 2 - 3 | 272 | 75 | |
GenBank mRNA | BC102961.1 | - | 2 - 3 | 275 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
