Assay Details
Gene Symbol: RAB19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035040.2 | NP_001030212.1 | 3 - 4 | 448 | 81 | |
GenBank mRNA | BC102961.1 | - | 3 - 4 | 451 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
