Assay Details
| Gene Symbol: SLC39A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035102.2 | NP_001030274.2 | 3 - 4 | 609 | 77 | |
| GenBank mRNA | BC102191.1 | - | 2 - 3 | 605 | 77 | |
| DV809336.1 | - | 3 - 4 | 611 | 77 | ||
| DV816868.1 | - | 2 - 3 | 423 | 77 | ||
| EV640746.1 | - | 3 - 4 | 668 | 77 | ||
More Information
Set Membership: |
|

SLC39A5
Genomic Map