Assay Details
Gene Symbol: RTRAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035280.1 | NP_001030357.1 | 4 - 5 | 444 | 126 | |
GenBank mRNA | BC102276.1 | - | 4 - 5 | 444 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
