Assay Details
| Gene Symbol: PTGR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035281.2 | NP_001030358.1 | 3 - 4 | 189 | 72 | |
| GenBank mRNA | BC102822.1 | - | 3 - 4 | 191 | 72 | |
More Information
Set Membership: |
|

PTGR1
Genomic Map