Assay Details
| Gene Symbol: SC5D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035356.1 | NP_001030433.1 | 5 - 6 | 641 | 91 | |
| GenBank mRNA | BC103338.1 | - | 5 - 6 | 641 | 91 | |
More Information
Set Membership: |
|

SC5D
Genomic Map