Assay Details
| Gene Symbol: AGT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114082.2 | NP_001107554.1 | 1 - 2 | 102 | 62 | |
| GenBank mRNA | BC102854.1 | - | 1 - 2 | 103 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102854.1, NM_001114082.2 transcripts.
Set Membership: |
|

AGT
Genomic Map