Assay Details
| Gene Symbol: AGT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114082.2 | NP_001107554.1 | 2 - 3 | 960 | 85 | |
| GenBank mRNA | BC102854.1 | - | 2 - 3 | 961 | 85 | |
More Information
Set Membership: |
|

AGT
Genomic Map