Assay Details
| Gene Symbol: ATP1B3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035393.2 | NP_001030470.1 | 4 - 5 | 694 | 90 | |
| GenBank mRNA | BC102454.1 | - | 4 - 5 | 696 | 90 | |
More Information
Set Membership: |
|

ATP1B3
Genomic Map