Assay Details
| Gene Symbol: BPGM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035402.2 | NP_001030479.1 | 2 - 3 | 767 | 72 | |
| GenBank mRNA | BC102611.1 | - | 2 - 3 | 768 | 72 | |
More Information
Set Membership: |
|

BPGM
Genomic Map