Assay Details
Gene Symbol: CNN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035420.1 | NP_001030497.1 | 2 - 3 | 240 | 77 | |
GenBank mRNA | BC103380.1 | - | 2 - 3 | 240 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
