Assay Details
Gene Symbol: BOLA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035452.2 | NP_001030529.1 | 2 - 3 | 179 | 103 | |
GenBank mRNA | BC103056.1 | - | 2 - 3 | 180 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
