Assay Details
| Gene Symbol: PRSS35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035457.3 | NP_001030534.3 | 1 - 2 | 169 | 110 | |
| GenBank mRNA | BC107557.1 | - | 1 - 2 | 157 | 110 | |
| BT020793.1 | - | 1 - 2 | 149 | 110 | ||
| DT807935.1 | - | 1 - 2 | 157 | 110 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC107557.1, NM_001035457.3 transcripts.
Set Membership: |
|

PRSS35
Genomic Map