Assay Details
Gene Symbol: BAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035459.2 | NP_001030536.1 | 3 - 4 | 442 | 62 | |
GenBank mRNA | BC103323.1 | - | 3 - 4 | 447 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
