Assay Details
| Gene Symbol: SAP18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035467.2 | NP_001030544.2 | 1 - 2 | 145 | 69 | |
| GenBank mRNA | BC102603.1 | - | 1 - 2 | 107 | 69 | |
| EV611947.1 | - | 1 - 2 | 147 | 69 | ||
More Information
Set Membership: |
|

SAP18
Genomic Map