Assay Details
| Gene Symbol: SAP18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035467.2 | NP_001030544.2 | 3 - 4 | 367 | 85 | |
| GenBank mRNA | BC102603.1 | - | 3 - 4 | 329 | 85 | |
| EE382620.1 | - | 1 - 2 | 64 | 85 | ||
| EV611947.1 | - | 3 - 4 | 369 | 85 | ||
More Information
Set Membership: |
|

SAP18
Genomic Map