Assay Details
Gene Symbol: CHMP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035472.1 | NP_001030549.1 | 2 - 3 | 321 | 95 | |
GenBank mRNA | BC104514.1 | - | 2 - 3 | 321 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
