Assay Details
| Gene Symbol: C15H11orf58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035474.2 | NP_001030551.1 | 1 - 2 | 182 | 102 | |
| GenBank mRNA | BC105190.1 | - | 1 - 2 | 183 | 102 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC105190.1, NM_001035474.2 transcripts.
Set Membership: |
|

C15H11orf58
Genomic Map