Assay Details
| Gene Symbol: FAM216A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035480.2 | NP_001030557.1 | 5 - 6 | 649 | 101 | |
| GenBank mRNA | BC102676.1 | - | 5 - 6 | 650 | 101 | |
More Information
Set Membership: |
|

FAM216A
Genomic Map