Assay Details
| Gene Symbol: RPH3AL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035494.1 | NP_001030571.1 | 7 - 8 | 784 | 66 | |
| GenBank mRNA | BC102095.1 | - | 7 - 8 | 784 | 66 | |
| BT021718.1 | - | 7 - 8 | 825 | 66 | ||
More Information
Set Membership: |
|

RPH3AL
Genomic Map