Assay Details
| Gene Symbol: UBE2V1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035497.1 | NP_001030574.1 | 1 - 2 | 44 | 75 | |
| GenBank mRNA | BC103134.1 | - | 1 - 2 | 44 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC103134.1, NM_001035497.1 transcripts.
Set Membership: |
|

UBE2V1
Genomic Map