Assay Details
| Gene Symbol: RPL3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035501.1 | NP_001030578.1 | 3 - 4 | 385 | 67 | |
| GenBank mRNA | BC103272.1 | - | 3 - 4 | 385 | 67 | |
More Information
Set Membership: |
|

RPL3L
Genomic Map