Assay Details
| Gene Symbol: NAT14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037245.1 | NP_001032322.1 | 1 - 2 | 515 | 61 | |
| GenBank mRNA | BC104515.2 | - | 1 - 2 | 515 | 61 | |
More Information
Set Membership: |
|

NAT14
Genomic Map