Assay Details
Gene Symbol: SLC16A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037319.1 | NP_001032396.1 | 1 - 2 | 147 | 131 | |
GenBank mRNA | BC104598.2 | - | 1 - 2 | 147 | 131 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC104598.2, NM_001037319.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
