Assay Details
| Gene Symbol: C1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037448.2 | NP_001032525.1 | 4 - 5 | 317 | 101 | |
| GenBank mRNA | BC108147.1 | - | 4 - 5 | 318 | 101 | |
More Information
Set Membership: |
|

C1D
Genomic Map