Assay Details
Gene Symbol: SLX1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037456.2 | NP_001032533.1 | 3 - 4 | 819 | 88 | |
GenBank mRNA | BC108108.1 | - | 3 - 4 | 820 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
