Assay Details
| Gene Symbol: SLX1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037456.2 | NP_001032533.1 | 5 - 6 | 1041 | 129 | |
| GenBank mRNA | BC108108.1 | - | 5 - 6 | 1042 | 129 | |
More Information
Set Membership: |
|

SLX1A
Genomic Map