Assay Details
| Gene Symbol: LEPROTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037462.2 | NP_001032539.1 | 3 - 4 | 378 | 96 | |
| GenBank mRNA | BC108155.1 | - | 3 - 4 | 379 | 96 | |
| BT025351.1 | - | 3 - 4 | 341 | 96 | ||
More Information
Set Membership: |
|

LEPROTL1
Genomic Map