Assay Details
| Gene Symbol: RPS19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037467.2 | NP_001032544.1 | 37 | 58 | ||
| GenBank mRNA | BC108158.1 | - | 38 | 58 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC108158.1, NM_001037467.2 transcripts.
Set Membership: |
|

RPS19
Genomic Map