Assay Details
| Gene Symbol: RPS19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037467.2 | NP_001032544.1 | 203 | 75 | ||
| GenBank mRNA | BC108158.1 | - | 204 | 75 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPS19| Gene Symbol: RPS19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037467.2 | NP_001032544.1 | 203 | 75 | ||
| GenBank mRNA | BC108158.1 | - | 204 | 75 | ||
Set Membership: |
|