Assay Details
Gene Symbol: RPS19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037467.2 | NP_001032544.1 | 442 | 145 | ||
GenBank mRNA | BC108158.1 | - | 443 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
