Assay Details
| Gene Symbol: APOA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037480.1 | NP_001032557.1 | 1 - 2 | 162 | 129 | |
| GenBank mRNA | BC108096.1 | - | 1 - 2 | 162 | 129 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC108096.1, NM_001037480.1 transcripts.
Set Membership: |
|

APOA4
Genomic Map