Assay Details
| Gene Symbol: APOA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037480.1 | NP_001032557.1 | 2 - 3 | 285 | 96 | |
| GenBank mRNA | BC108096.1 | - | 2 - 3 | 285 | 96 | |
More Information
Set Membership: |
|

APOA4
Genomic Map