Assay Details
| Gene Symbol: LSM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037487.2 | NP_001032564.1 | 1 - 2 | 37 | 54 | |
| GenBank mRNA | BC108142.1 | - | 1 - 2 | 38 | 54 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC108142.1, NM_001037487.2 transcripts.
Set Membership: |
|

LSM3
Genomic Map