Assay Details
| Gene Symbol: TMEM205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037599.2 | NP_001032688.1 | 3 - 4 | 391 | 84 | |
| GenBank mRNA | BC109820.1 | - | 3 - 4 | 392 | 84 | |
More Information
Set Membership: |
|

TMEM205
Genomic Map