Assay Details
| Gene Symbol: MRPS27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038108.2 | NP_001033197.1 | 7 - 8 | 601 | 59 | |
| GenBank mRNA | BC108100.1 | - | 7 - 8 | 602 | 59 | |
More Information
Set Membership: |
|

MRPS27
Genomic Map