Assay Details
| Gene Symbol: MYCBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038565.2 | NP_001033654.1 | 4 - 5 | 276 | 105 | |
| GenBank mRNA | BC109848.1 | - | 4 - 5 | 277 | 105 | |
More Information
Set Membership: |
|

MYCBP
Genomic Map