Assay Details
Gene Symbol: RPL19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040516.2 | NP_001035606.1 | 3 - 4 | 272 | 60 | |
GenBank mRNA | BC102223.1 | - | 3 - 4 | 273 | 60 | |
BC142030.1 | - | 3 - 4 | 270 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
