Assay Details
| Gene Symbol: RPS21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040581.2 | NP_001035671.1 | 287 | 56 | ||
| GenBank mRNA | BC108180.1 | - | 288 | 56 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPS21| Gene Symbol: RPS21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040581.2 | NP_001035671.1 | 287 | 56 | ||
| GenBank mRNA | BC108180.1 | - | 288 | 56 | ||
Set Membership: |
|