Assay Details
| Gene Symbol: COX20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040585.2 | NP_001035675.1 | 2 - 3 | 271 | 64 | |
| GenBank mRNA | BC109923.1 | - | 2 - 3 | 272 | 64 | |
More Information
Set Membership: |
|

COX20
Genomic Map